Anti-CENPF Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309605EA 8950 NOK
ORIGTA309605
Anti-CENPF Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: CENPF (centromere protein F, 350/400kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR