Anti-DCBLD2 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309619EA 6400 NOK
ORIGTA309619
Anti-DCBLD2 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: DCBLD2 (discoidin, CUB and LCCL domain containing 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR