Anti-DDIT3 Mouse Monoclonal Antibody [clone: 9C8]

Leverandør: OriGene
ORIGTA309621EA 8500 NOK
ORIGTA309621
Anti-DDIT3 Mouse Monoclonal Antibody [clone: 9C8]
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: DDIT3 (DNA-damage-inducible transcript 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR