Anti-DEFA5 Mouse Monoclonal Antibody [clone: 8C8]

Leverandør: OriGene
ORIGTA309622EA 7850 NOK
ORIGTA309622
Anti-DEFA5 Mouse Monoclonal Antibody [clone: 8C8]
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: DEFA5 (defensin, alpha 5, Paneth cell-specific)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR