Anti-EGLN3 Mouse Monoclonal Antibody [clone: EG188e/d5]

Leverandør: OriGene
ORIGTA309636EA 7850 NOK
ORIGTA309636
Anti-EGLN3 Mouse Monoclonal Antibody [clone: EG188e/d5]
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: EGLN3 (egl-9 family hypoxia-inducible factor 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR