Anti-EPB41L3 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309642EA 7475 NOK
ORIGTA309642
Anti-EPB41L3 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: EPB41L3 (erythrocyte membrane protein band 4.1-like 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR