Anti-ESPL1 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309645EA 7475 NOK
ORIGTA309645
Anti-ESPL1 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: ESPL1 (extra spindle pole bodies homolog 1 (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR