Anti-ESPL1 Mouse Monoclonal Antibody [clone: XJ11-1B12]

Leverandør: OriGene
ORIGTA309646EA 7475 NOK
ORIGTA309646
Anti-ESPL1 Mouse Monoclonal Antibody [clone: XJ11-1B12]
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: ESPL1 (extra spindle pole bodies homolog 1 (S. cerevisiae))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR