Anti-F2R Mouse Monoclonal Antibody [clone: N2-11]

Leverandør: OriGene
ORIGTA309650EA 7850 NOK
ORIGTA309650
Anti-F2R Mouse Monoclonal Antibody [clone: N2-11]
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: F2R (coagulation factor II (thrombin) receptor)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR