Anti-PIEZO2 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309653EA 8500 NOK
ORIGTA309653
Anti-PIEZO2 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: PIEZO2 (piezo-type mechanosensitive ion channel component 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR