Anti-FANCM Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309657EA 6775 NOK
ORIGTA309657
Anti-FANCM Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: FANCM (Fanconi anemia, complementation group M)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR