Anti-FBXO17 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309659EA 6400 NOK
ORIGTA309659
Anti-FBXO17 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: FBXO17 (F-box protein 17)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR