Anti-FXR2 Mouse Monoclonal Antibody [clone: 1G2]

Leverandør: OriGene
ORIGTA309666EA 7475 NOK
ORIGTA309666
Anti-FXR2 Mouse Monoclonal Antibody [clone: 1G2]
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: FXR2 (fragile X mental retardation, autosomal homolog 2)
Clonality: Monoclonal
Clone: 1G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR