Anti-GLO1 Mouse Monoclonal Antibody [clone: Glo1a]

Leverandør: OriGene
ORIGTA309675EA 7850 NOK
ORIGTA309675
Anti-GLO1 Mouse Monoclonal Antibody [clone: Glo1a]
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: GLO1 (glyoxalase I)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR