Anti-HIF1A Mouse Monoclonal Antibody [clone: ESEE122]

Leverandør: OriGene
ORIGTA309690EA 8950 NOK
ORIGTA309690
Anti-HIF1A Mouse Monoclonal Antibody [clone: ESEE122]
Antistoffer
  • Recommended Dilutions:

Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR