Anti-HIP1 Mouse Monoclonal Antibody [clone: 1B11]

Leverandør: OriGene
ORIGTA309694EA 7475 NOK
ORIGTA309694
Anti-HIP1 Mouse Monoclonal Antibody [clone: 1B11]
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: HIP1 (huntingtin interacting protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR