Anti-IGF1R Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309698EA 7475 NOK
ORIGTA309698
Anti-IGF1R Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: IGF1R (insulin-like growth factor 1 receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR