Anti-KCNA2 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309707EA 7475 NOK
ORIGTA309707
Anti-KCNA2 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: KCNA2 (potassium voltage-gated channel, shaker-related subfamily, member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR