Anti-KDM4D Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309712EA 6775 NOK
ORIGTA309712
Anti-KDM4D Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: KDM4D (lysine (K)-specific demethylase 4D)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR