Anti-LDLR Mouse Monoclonal Antibody [clone: C7]
Leverandør: OriGene
ORIGTA309729EA 7850 NOK
ORIGTA309729
Anti-LDLR Mouse Monoclonal Antibody [clone: C7]
Antistoffer
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100; Immunochemistry: 1:200; Immunoprecipitation: 1:10-1:500
Type: Primary
Antigen: LDLR (low density lipoprotein receptor)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestill nå