Anti-LRRK2 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309735EA 7475 NOK
ORIGTA309735
Anti-LRRK2 Rabbit Polyclonal Antibody
Antistoffer
  • Recommended Dilutions:

Type: Primary
Antigen: LRRK2 (leucine-rich repeat kinase 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR