Anti-MAP2 Mouse Monoclonal Antibody [clone: 5H11]
Leverandør: OriGene
ORIGTA309740EA 7850 NOK
ORIGTA309740
Anti-MAP2 Mouse Monoclonal Antibody [clone: 5H11]
Antistoffer
Recommended Dilutions:
Type: Primary
Antigen: MAP2 (microtubule-associated protein 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 Kappa
Reactivity: Bovine, Human, Mouse, Rat