Anti-NBR1 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309759EA 7850 NOK
ORIGTA309759
Anti-NBR1 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: NBR1 (neighbor of BRCA1 gene 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR