Anti-NPC1L1 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309769EA 7350 NOK
ORIGTA309769
Anti-NPC1L1 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: NPC1L1 (NPC1-like 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR