Anti-OPRK1 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309784EA 6400 NOK
ORIGTA309784
Anti-OPRK1 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: OPRK1 (opioid receptor, kappa 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR