Anti-PALLD Mouse Monoclonal Antibody [clone: 1E6]

Leverandør: OriGene
ORIGTA309787EA 7850 NOK
ORIGTA309787
Anti-PALLD Mouse Monoclonal Antibody [clone: 1E6]
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: PALLD (palladin, cytoskeletal associated protein)
Clonality: Monoclonal
Clone: 1E6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR