Anti-PCNA Mouse Monoclonal Antibody [clone: PC10]

Leverandør: OriGene
ORIGTA309795EA 7700 NOK
ORIGTA309795
Anti-PCNA Mouse Monoclonal Antibody [clone: PC10]
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: PCNA (proliferating cell nuclear antigen)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR