Anti-PODXL Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309811EA 7850 NOK
ORIGTA309811
Anti-PODXL Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: PODXL (podocalyxin-like)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR