Anti-PPP2R5C Mouse Monoclonal Antibody [clone: TQ11-1G6]

Leverandør: OriGene
ORIGTA309817EA 5775 NOK
ORIGTA309817
Anti-PPP2R5C Mouse Monoclonal Antibody [clone: TQ11-1G6]
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: PPP2R5C (protein phosphatase 2, regulatory subunit B', gamma)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR