Anti-RETN Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309836EA 7475 NOK
ORIGTA309836
Anti-RETN Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: RETN (resistin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR