Anti-SETX Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309853EA 7850 NOK
ORIGTA309853
Anti-SETX Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:500; Immunoprecipitation: 1:500; Immunofluorescence/Immunochemistry: 1:100

Type: Primary
Antigen: SETX (senataxin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR