Anti-SF3B4 Mouse Monoclonal Antibody [clone: 3A1]

Leverandør: OriGene
ORIGTA309854EA 7475 NOK
ORIGTA309854
Anti-SF3B4 Mouse Monoclonal Antibody [clone: 3A1]
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: SF3B4 (splicing factor 3b, subunit 4, 49kDa)
Clonality: Monoclonal
Clone: 3A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR