Anti-SLC45A3 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309870EA 7850 NOK
ORIGTA309870
Anti-SLC45A3 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: SLC45A3 (solute carrier family 45, member 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR