Anti-STRN3 Mouse Monoclonal Antibody [clone: S68]

Leverandør: OriGene
ORIGTA309886EA 7850 NOK
ORIGTA309886
Anti-STRN3 Mouse Monoclonal Antibody [clone: S68]
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: STRN3 (striatin, calmodulin binding protein 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR