Anti-TERF1 Mouse Monoclonal Antibody [clone: 57-6]

Leverandør: OriGene
ORIGTA309899EA 7850 NOK
ORIGTA309899
Anti-TERF1 Mouse Monoclonal Antibody [clone: 57-6]
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: TERF1 (telomeric repeat binding factor (NIMA-interacting) 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR