Anti-TET1 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309902EA 7475 NOK
ORIGTA309902
Anti-TET1 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Immunochemistry: 1:400

Type: Primary
Antigen: TET1 (tet methylcytosine dioxygenase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR