Anti-UBIAD1 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309925EA 6400 NOK
ORIGTA309925
Anti-UBIAD1 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: UBIAD1 (UbiA prenyltransferase domain containing 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR