Anti-WNK4 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309931EA 7850 NOK
ORIGTA309931
Anti-WNK4 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: WNK4 (WNK lysine deficient protein kinase 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR