Anti-ACAT1 Mouse Monoclonal Antibody [clone: AT1.H11]

Leverandør: OriGene
ORIGTA309940EA 6675 NOK
ORIGTA309940
Anti-ACAT1 Mouse Monoclonal Antibody [clone: AT1.H11]
Antistoffer
Recommended Dilutions: Western Blot: 1:1000; ELISA: 1:2000-1:5000

Type: Primary
Antigen: ACAT1 (acetyl-CoA acetyltransferase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR