Anti-STAT3 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309956EA 8650 NOK
ORIGTA309956
Anti-STAT3 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: STAT3 (signal transducer and activator of transcription 3 (acute-phase response factor))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR