Anti-ATF4 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309962EA 8650 NOK
ORIGTA309962
Anti-ATF4 Rabbit Polyclonal Antibody
Antistoffer
  • Recommended Dilutions: Immunochemistry: 1:50-1:100

Type: Primary
Antigen: ATF4 (activating transcription factor 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR