Anti-HDAC8 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309982EA 8650 NOK
ORIGTA309982
Anti-HDAC8 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Immunochemistry: 1:50-1:100

Type: Primary
Antigen: HDAC8 (histone deacetylase 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR