Anti-HDAC4 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA310003EA 8650 NOK
ORIGTA310003
Anti-HDAC4 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: HDAC4 (histone deacetylase 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR