Anti-SMAD1 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA310019EA 8650 NOK
ORIGTA310019
Anti-SMAD1 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: SMAD1 (SMAD family member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR