Anti-SMAD2 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA310020EA 8650 NOK
ORIGTA310020
Anti-SMAD2 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: SMAD2 (SMAD family member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR