Anti-PTPRC Rat Monoclonal Antibody

Leverandør: OriGene
ORIGTA310023EA 6675 NOK
ORIGTA310023
Anti-PTPRC Rat Monoclonal Antibody
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR