Anti-ITGA5 Rat Monoclonal Antibody

ORIGTA310024EA 5150 NOK
ORIGTA310024
Anti-ITGA5 Rat Monoclonal Antibody
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: ITGA5 (integrin, alpha 5 (fibronectin receptor, alpha polypeptide))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR