Anti-ACAT2 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA310033EA 4600 NOK
ORIGTA310033
Anti-ACAT2 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: ACAT2 (acetyl-CoA acetyltransferase 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR