Anti-ACAT1 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA310034EA 4600 NOK
ORIGTA310034
Anti-ACAT1 Rabbit Polyclonal Antibody
Antistoffer
  • Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: ACAT1 (acetyl-CoA acetyltransferase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR