Anti-CAPG Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA310036EA 5425 NOK
ORIGTA310036
Anti-CAPG Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:250

Type: Primary
Antigen: CAPG (capping protein (actin filament), gelsolin-like)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR